ESD occupational shoes, sandals with Velcro®, Hand-sewn, Traunreut, 52142
Supplier: SCHURR SCHUHVERTRIEB
5.2142.01.38 5.2142.01.36 5.2142.01.37 5.2142.01.39 5.2142.01.40 5.2142.01.41 5.2142.01.42 5.2142.01.43 5.2142.01.44 5.2142.01.45 5.2142.01.46 5.2142.01.47 5.2142.01.48
SCRR5.2142.01.38EA 0 EUR
SCRR5.2142.01.38 SCRR5.2142.01.36 SCRR5.2142.01.37 SCRR5.2142.01.39 SCRR5.2142.01.40 SCRR5.2142.01.41 SCRR5.2142.01.42 SCRR5.2142.01.43 SCRR5.2142.01.44 SCRR5.2142.01.45 SCRR5.2142.01.46 SCRR5.2142.01.47 SCRR5.2142.01.48
ESD occupational shoes, sandals with Velcro®, Hand-sewn, Traunreut, 52142
Shoes and Boots
Comfortable hand-sewn ESD occupational shoe made from smooth leather, DRYtech® lining and a PU sole; with adjustable heel strap.
  • Antibacterial and breathable DRYtech® textile lining
  • Anatomically shaped removable PUR-foam insole prevents from fatigue
  • Microfibre reinforced heel
  • High-quality hand seam makes shoe soft and flexible
For ESD and kitchen use.
Colour: White
Width: 9
Weight: 360 g
Certifications: EN ISO 20347:2012; OB