Ethyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
H35916.03 H35916.06
H35916.03EA 63.1 EUR
H35916.03 H35916.06
Ethyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₁H₈ClF₃N₂O₂
MW: 292.65 g/mol
Melting Pt: 132…136 °C
Storage Temperature: Ambient
MDL Number: MFCD01833013
CAS Number: 353258-31-8