Diethylzinc 15% ∼1.1 mol/l in toluene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
220809-100G 220809-800G
SIAL220809-100GEA 178 EUR
SIAL220809-100G SIAL220809-800G
Diethylzinc 15% ∼1.1 mol/l in toluene, Sigma-Aldrich®
Diethylzinc
Formula: C₄H₁₀Zn
MDL Number: MFCD00009021
CAS Number: 557-20-0
EINECS: 209-161-3