Tin(II) tetrafluoroborate 50% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Danger
89669.A1 89669.D5
89669.A1EA 147 EUR
89669.A1 89669.D5
Tin(II) tetrafluoroborate 50% (w/w) in aqueous solution
Tin (II) tetrafluoroborate
CAS No.: 13814-97-6 Molecular Formula: B2F8Sn Formula Weight: 292.30 Physical Form: Liquid Density: 1.670 Appearance: Colourless UN No.: 3264 MDL No.: MFCD00054047
Formula: B₂F₈Sn
MW: 292.30 g/mol
Density: 1.670
Storage Temperature: Ambient
MDL Number: MFCD00054047
CAS Number: 13814-97-6
EINECS: 237-487-6
UN: 3264
ADR: 8 III