4-Cyanobenzamide ≥97%

Supplier: Thermo Fisher Scientific
Danger

B25600.03 B25600.06
B25600.03EA 57.4 EUR
B25600.03 B25600.06
4-Cyanobenzamide ≥97%
4-Cyanobenzamide

Purity: 97% CAS No.: 3034-34-2 Molecular Formula: C8H6N2O Formula Weight: 146.15 Melting Point: 225-228° MDL No.: MFCD00017133

Formula: C₈H₆N₂O
Melting Pt: 225…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00017133
CAS Number: 3034-34-2
EINECS: 221-223-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR