4-Iodo-5-methyl-3-phenylisoxazole

Supplier: Thermo Scientific

CC00511DA CC00511EA
MAYBCC00511DAEA 0 EUR
MAYBCC00511DA MAYBCC00511EA
4-Iodo-5-methyl-3-phenylisoxazole
4-Iodo-5-methyl-3-phenylisoxazole
Formula: C₁₀H₈INO
MDL Number: MFCD02681888
CAS Number: 31295-66-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR