2,2':6',2''-Terpyridine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
234672-1G 234672-250MG 234672-5G
SIAL234672-1GEA 151 EUR
SIAL234672-1G SIAL234672-250MG SIAL234672-5G
2,2':6',2''-Terpyridine, Sigma-Aldrich®
2,2':6',2''-Terpyridine
Formula: C₁₅H₁₁N₃
MDL Number: MFCD00006213
CAS Number: 1148-79-4
EINECS: 214-559-5