Hydrogen chloride 1.25 M in ethanol, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
427011000 427018000 427011000_T 427018000_T
427011000.EA 50.4 EUR
427011000. 427018000. 427011000_T 427018000_T
Hydrogen chloride 1.25 M in ethanol, AcroSeal®
Hydrogen chloride
Formula: HCl
MW: 36.46 g/mol
Flash Pt: 12 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless Solution
Water ≤0.5 %
Strength 1.10 to 1.40 M
NMR ≥96.0 %