Dimethylaluminium chloride 0.9 M in heptane, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
377791000 377798000
377791000.EA 88 EUR
377791000. 377798000.
Dimethylaluminium chloride 0.9 M in heptane, AcroSeal®
Dimethylaluminium chloride
Formula: C₂H₆AlCl
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000458
CAS Number: 1184-58-3
EINECS: 214-668-8
UN: 3399
ADR: 4.3,I
Order Now
Specification Test Results
Appearance Clear to slightly hazy colorless Solution (May develop some turbidity or precipitate)
Titration Argentometric 0.85 to 1.00 M