TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water
Supplier: G Bioscienes
Danger
Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
DG907 DG908
DG907EA 335 EUR
DG907 DG908
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 6
Density: ∼1.01 g/cm³ (20 °C)
Flash Pt: 251
Storage Temperature: Ambient
MDL Number: MFCD00132505
CAS Number: 9002-93-1
Merck Index: 13,06793