Tween® 20 (Polysorbate) 10% in aqueous solution, Proteomics Grade
Supplier: G Bioscienes
Synonyms: Polyoxyethylene sorbitan monolaurate
DG011 DG012
DG011EA 289 EUR
DG011 DG012
Tween® 20 (Polysorbate) 10% in aqueous solution, Proteomics Grade
Tween® 20 (Polysorbate)
Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3