Sodium chloride 5 mol/l (5 N) in aqueous solution
Supplier: Cayman Chemical
Synonyms: Saline, NaCl, Sea salt, Halite
600211-100 600211-250 600211-500
CAYM600211-100EA 27 EUR
CAYM600211-100 CAYM600211-250 CAYM600211-500
Sodium chloride 5 mol/l (5 N) in aqueous solution
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599