Sulphuric acid ≥92% (metals basis)
Supplier: Thermo Fisher Scientific
Danger
11000.AB 11000.AE 11000.AP
11000.ABEA 60.5 EUR
11000.AB 11000.AE 11000.AP
Sulphuric acid ≥92% (metals basis)
Sulphuric acid
Purity: 99.9999% CAS No.: 7664-93-9 Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid Density: 1.84 Appearance: Colourless to pale yellow Odour: Acrid Merck Reference: 14,8974 UN No.: 1830 MDL No.: MFCD00064589
Formula: H₂SO₄
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,8974