Sodium hydroxide 1 mol/l (1 N) in aqueous solution 1 mol/l (1 N) in aqueous solution
Supplier: Hach
Danger
Synonyms: Caustic soda
104532 104542
HACH104532EA 24.7 EUR
HACH104532 HACH104542
Sodium hydroxide 1 mol/l (1 N) in aqueous solution 1 mol/l (1 N) in aqueous solution
Sodium hydroxide
Formula: NaOH
Density: 1.04 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 3266
ADR: 8,III