Aluminium chloride standardised solution 6.0 N in aqueous solution
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Aluminum chloride
35568.AE 35568.AK
35568.AEEA 80.1 EUR
35568.AE 35568.AK
Aluminium chloride standardised solution 6.0 N in aqueous solution
Aluminium chloride
Molecular Formula: AlCl3 Formula Weight: 133.34 Physical Form: Liquid Odour: Odourless UN No.: 2581 MDL No.: MFCD00003422
Formula: AlCl₃
MW: 133.34 g/mol
Boiling Pt: 180
Storage Temperature: Ambient
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 2581
ADR: 8 III
Merck Index: 13,00335