ESD occupational shoes, sandals, Tiefbett Leder, Bad Tölz, 51938
Supplier: SCHURR SCHUHVERTRIEB
5.1938.04.42 5.1938.04.46 5.1938.04.43
SCRR5.1938.04.42EA 63.1 EUR
SCRR5.1938.04.42 SCRR5.1938.04.46 SCRR5.1938.04.43
ESD occupational shoes, sandals, Tiefbett Leder, Bad Tölz, 51938
Shoes and Boots
Deeply moulded occupational shoes made from perforated leather and an anti-slip and anti-static 'A4' nitrile rubber sole; with adjustable heel strap.
  • Anatomically shaped stabilising integrated insole made from cork/latex
  • Insole features cushioned heel, ankle support and toe bar
  • Shock absorber heel
For ESD and kitchen use.
Colour: Blue
Width: 9
Weight: 295 g
Certifications: EN 61340 (ESD); EN ISO 20347:2012; OB, SRC