3-Pyridineacetamide ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
B25692.14
B25692.06
B25692.14EA
207
EUR
B25692.14
B25692.06
3-Pyridineacetamide ≥99%
3-Pyridineacetamide
Purity: 99%
CAS No.: 3724-16-1
Molecular Formula: C7H8N2O
Formula Weight: 136.15
Melting Point: 122-124°
MDL No.: MFCD08236789
Formula:
C₇H₈N₂O Melting Pt: 122…124 °C Density: 1.17 Storage Temperature: Ambient |
MDL Number:
MFCD08236789 CAS Number: 3724-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...