(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, Sigma-Aldrich®
Supplier: Merck
Synonyms: (S)-(+)-1,1'-Binaphthalene-2,2'-diyl-phosphate, (11bS)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide
M-Clarity™ quality level = MQ100
248940-1G 248940-5G
SIAL248940-1GEA 108 EUR
SIAL248940-1G SIAL248940-5G
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, Sigma-Aldrich®
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
Formula: C₂₀H₁₃O₄P
MW: 348.289 g/mol
MDL Number: MFCD00010045
CAS Number: 35193-64-7