2-Mercaptothiophene, Sigma-Aldrich®
Supplier: Merck
Synonyms: Thiophene-2-thiol
M-Clarity™ quality level = MQ300
W306207-100G-K W306207-1KG-K
SIALW306207-100G-KEA 568 EUR
SIALW306207-100G-K SIALW306207-1KG-K
2-Mercaptothiophene, Sigma-Aldrich®
2-Mercaptothiophene
Formula: C₄H₄S₂
MDL Number: MFCD00051666
CAS Number: 7774-74-5
EINECS: 231-881-1