2-Methylindene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

449431-1G 449431-5G
SIAL449431-1GEA 81.8 EUR
SIAL449431-1G SIAL449431-5G
2-Methylindene, Sigma-Aldrich®
2-Methylindene
Formula: C₁₀H₁₀
MDL Number: MFCD00274253
CAS Number: 2177-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR