Capillary columns, Rtx®-200
Supplier: Restek
45001 45002 45003 45010 45011 45012 15005 15006 15007 15008 15009 15010 15020 15021 15022 15023 15024 15025 15026 15027 15028 15029 15035 15036 15037 15038 15039 15040 15041 15042 15043 15044 15045 15050 15051 15052 15053 15054 15055 15056 15057 15058 15059 15060 15066 15067 15069 15070 15072 15073 15075 15082 15085 15088 15091 43835 43836
568-2473EA 437 EUR
568-2473 568-2474 568-2475 568-2476 568-2477 568-2478 568-2242 568-2243 568-2244 568-2245 568-2246 568-2247 568-2251 568-2252 568-2253 568-2254 568-2255 568-2256 568-2257 568-2258 568-2259 568-2260 568-2262 568-2263 568-2264 568-2265 568-2266 568-2267 568-2268 568-2269 568-2270 568-2271 568-2272 568-2273 568-2274 568-2275 568-2276 568-2277 568-2278 568-2279 568-2280 568-2281 568-2282 568-2283 568-2284 568-2285 568-2286 568-2287 568-2288 568-2289 568-2290 568-2291 568-2292 568-2293 568-2294 REST43835 REST43836
Capillary columns, Rtx®-200
Chromatography Columns
General purpose columns for solvents, Freon® fluorocarbons, alcohols, ketones, silanes, glycols and drugs of abuse. Excellent confirmation column with an Rtx®-5 column for phenols, nitrosamines, organochlorine pesticides, chlorinated hydrocarbons and chlorophenoxy herbicides.
Midpolarity phase; Crossbond® trifluoropropylmethyl polysiloxane
Temperature range: –20 °C to 340 °C
Suitable replacement for: DB-200, DB-210. Equivalent to USP G6 phase.