Drum stands, FB
Supplier: LACONT UMWELTTECHNIK
P21-1701-A P21-1702-A P21-1703-A P21-1802-A P21-1801-A P21-1803-A
LACOP21-1701-AEA 0 EUR
LACOP21-1701-A LACOP21-1702-A LACOP21-1703-A LACOP21-1802-A LACOP21-1801-A LACOP21-1803-A
Drum stands, FB
Drum Storage
Drum stands with fixed drum supports for 60 to 200 L drums, completely galvanised. Tilted support onwards for easy drum emptying.
These drum stands can be combined with compact sump pallets, water protection pallets, hazardous substance stations, floor safety elements and drum carts.