Sumps for KTC/IBC storage, PE-KT-EP
Supplier: LACONT UMWELTTECHNIK
P70-7100-A P70-7102-A P70-7104-A P70-7105-A P70-7103-A P70-7101-A P70-7134-A P70-7133-A P70-7135-A P70-7136-A
LACOP70-7100-AEA 0 EUR
LACOP70-7100-A LACOP70-7102-A LACOP70-7104-A LACOP70-7105-A LACOP70-7103-A LACOP70-7101-A LACOP70-7134-A LACOP70-7133-A LACOP70-7135-A LACOP70-7136-A
Sumps for KTC/IBC storage, PE-KT-EP
Drum Storage
Sumps are used for storing water-endangering and aggressive media.
  • Rotationally-molded body of black PE
  • Circumferential stabilising ribs ensure high load values
  • Ground clearance for internal transport
  • Galvanised or PE grid support
  • Cutouts for easily removable grid