Folding steps
Supplier: GUENZBURGER STEIGTECHNIK
55002 54202 54203
139-1020EA 0 EUR
139-1020 GUEN54202 139-0770
Folding steps
Ladders and Step Stools
Foldable steps, sturdy double side rails, grooved aluminium steps (205×360 mm).
  • Easy fold away
  • Non slip and robust for a safe working
  • Measures only 85 mm in depth when folded
Certifications: GS tested; in accordance with European standard DIN EN 14183.
Action manual BGI 694 and the applicable accident prevention regulations.