Extraction fans for storeLAB®
Supplier: LACONT UMWELTTECHNIK
B80-2839-A B80-2841-A B80-2842-A B80-2844-A B80-2846-A B80-2843-A B80-2847-A
LACOB80-2839-AEA 709 EUR
LACOB80-2839-A LACOB80-2841-A LACOB80-2842-A LACOB80-2844-A LACOB80-2846-A LACOB80-2843-A LACOB80-2847-A
Extraction fans for storeLAB®
Ventilators
Safety cabinets for the storage of hazardous substances are provided with a DN 80 exhaust air connection. The forced ventilation can be carried out by one of our standard extraction fans or by a suitable in-house air extraction system. The exhaust air is to be discharged from the building into a hazard-free zone.
  • Mounting plate for easy mounting on cabinet roof
  • When operating the cabinets without technical forced ventilation, additional replacement measures have to be taken to ensure comparable explosion protection
  • To avoid the presence of hazardous vapours in the atmosphere near the cabinet safety cabinets are operated with an extraction fan for proper operation
Certifications: DIN EN 14470-1, TRGS 510, EU Directive 94/9/EU (ATEX).