Capillary columns, TraceGOLD™ TG-5MS
Supplier: Thermo Fisher Scientific
26098-2860 26098-1550 26098-2130 26098-2230 26098-2240 26098-2250 26098-2350 26098-2360 26098-1440 26098-2850 26098-1430 26098-2960 26098-2970 26098-2980 26098-3080 26098-3090 26098-3340 26098-3360 26098-5780 26098-1540 26098-0200 26098-1300 26098-1310 26098-1420 26098-2760 26098-2940
555-9348EA 618 EUR
555-9348 555-8680 555-8677 555-8676 555-8674 555-8673 555-8672 555-8671 555-8682 555-9005 555-8684 555-9346 555-9345 555-9344 555-9343 555-9342 555-9341 555-9340 555-9337 555-8681 555-8692 555-8690 555-8688 555-8686 555-8669 555-9347
Capillary columns, TraceGOLD™ TG-5MS
Chromatography Columns
Employ the most widely used MS phase in gas chromatography with the 5% phenyl phase Thermo Scientific TraceGOLD TG-5MS GC Column. These columns have exceptionally low bleed for optimal signal-to-noise ratio, sensitivity and MS integrity.
  • TG-5MS Fast increase speed of analysis by a factor of 3 to 10 times
  • Low polarity phase, 5% diphenyl/95% dimethylm polysiloxane
  • High thermal stability - maximum temperatures up to 330/350 °C
  • Ideal for analysis of active compounds
Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity. Exceptional inertness ideal for analysis of active compounds. Equivalent to USP G27 phase. Similar to Rxi-5ms, DB-5, HP-5, HP-5ms, Ultra-2, SPB-5, Equity-5, CP-Sil 8. Recommended for semivolatiles, phenols, amines, residual solvents and solvent impurities, drugs of abuse, pesticides, PCB cogeners, and aroclor mixes.