Filter tips, epT.I.P.S.® Motion
Supplier: EPPENDORF
0030014.413 0030014.430 0030014.456 0030014.472 0030014.499 0030014.510 0030015.215 0030015.231 0030015.258 0030014.529 0030014.537 0030014.553 0030014.561 0030014.570
EPPE0030014.413EA 228 EUR
EPPE0030014.413 EPPE0030014.430 EPPE0030014.456 EPPE0030014.472 EPPE0030014.499 EPPE0030014.510 EPPE0030015.215 EPPE0030015.231 EPPE0030015.258 EPPE0030014.529 EPPE0030014.537 EPPE0030014.553 EPPE0030014.561 EPPE0030014.570
Filter tips, epT.I.P.S.® Motion
Pipette Tips
These filter tips are specially designed for use on epMotion Liquid Handling workstations. They were developed in torsion-resistant, contamination-free racks.
  • High-precision manufacturing guarantees a perfect fit and extremely accurate dispensing behavior
  • Tips are optimised for simple automatic ejection in the Liquid Handling Workstation collection vessel
  • Tips can also be integrated into the workstation as a refilling system
  • Refilling tips are delivered in a PET thermoformed tray with sealed lid
  • Replacing the disposable box with an autoclavable aluminum adapter can mean a waste savings of up to 40 percent
Motion pipette tips are available in the following purity levels: PCR clean and PCR sterile+clean.
The epT.I.P.S. Motion Rack is made up of two components for flexible use in the automated epMotion Liquid Handling Workstation: a colour tray with the epT.I.P.S. filter tips and a disposable box for storing the tray.
Packaging: Packaged in contamination-free racks or in reload trays.