ESD ocupational shoes, clogs, Dynamic, 37310 to 37315
Supplier: Abeba
37315/37 37312/47 37315/35 37315/48 37313/47 37313/46 37313/45 37313/44 37313/43 37313/42 37313/41 37313/40 37313/39 37313/38 37313/37 37313/36 37315/36 37310/48 37310/35 37312/46 37312/45 37312/44 37312/43 37312/42 37312/41 37312/40 37312/39 37312/38 37312/37 37312/48 37312/35 37313/35 37310/47 37310/46 37310/45 37310/44 37310/43 37310/42 37310/41 37310/40 37310/39 37310/38 37310/37 37310/36 37312/36 37313/48 37315/47 37315/46 37315/45 37315/44 37315/43 37315/42 37315/41 37315/40 37315/39 37315/38
ABEB37315/37EA 61.2 EUR
ABEB37315/37 ABEB37312/47 ABEB37315/35 ABEB37315/48 ABEB37313/47 ABEB37313/46 ABEB37313/45 ABEB37313/44 ABEB37313/43 ABEB37313/42 ABEB37313/41 ABEB37313/40 ABEB37313/39 ABEB37313/38 ABEB37313/37 ABEB37313/36 ABEB37315/36 ABEB37310/48 ABEB37310/35 ABEB37312/46 ABEB37312/45 ABEB37312/44 ABEB37312/43 ABEB37312/42 ABEB37312/41 ABEB37312/40 ABEB37312/39 ABEB37312/38 ABEB37312/37 ABEB37312/48 ABEB37312/35 ABEB37313/35 ABEB37310/47 ABEB37310/46 ABEB37310/45 ABEB37310/44 ABEB37310/43 ABEB37310/42 ABEB37310/41 ABEB37310/40 ABEB37310/39 ABEB37310/38 ABEB37310/37 ABEB37310/36 ABEB37312/36 ABEB37313/48 ABEB37315/47 ABEB37315/46 ABEB37315/45 ABEB37315/44 ABEB37315/43 ABEB37315/42 ABEB37315/41 ABEB37315/40 ABEB37315/39 ABEB37315/38
ESD ocupational shoes, clogs, Dynamic, 37310 to 37315
Shoes and Boots
ESD clog made from smooth leather with a foldable and adjustable heel strap. The TPU sole is resistant to fuels, oils and animal fats.
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • TPU sole with high slip resistance
Colour: White, white/blue, white/red, black/brown
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; OB, FO,E,A, SRC.