3,6-Dibromopyridazine 95%
Supplier: Thermo Fisher Scientific
Warning
H61223.03 H61223.06
H61223.03EA 68.5 EUR
H61223.03 H61223.06
3,6-Dibromopyridazine 95%
3,6-Dibromopyridazine
Formula: C₄H₂Br₂N₂
MW: 237.88 g/mol
Melting Pt: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00233947
CAS Number: 17973-86-3