Tantalum nitride ≥99.5% (metals basis)
Supplier: Thermo Fisher Scientific
13093.09
13093.18
13093.09EA
85.3
EUR
13093.09
13093.18
Tantalum nitride ≥99.5% (metals basis)
Tantalum nitride
Purity: 99.5%
CAS No.: 12033-62-4
Molecular Formula: TaN
Formula Weight: 194.95
Physical Form: -325 Mesh Powder
Odour: Odourless
MDL No.: MFCD00049568
Formula:
NTa MW: 194.95 g/mol Melting Pt: 3360 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049568 CAS Number: 12033-62-4 EINECS: 234-788-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...