S-Furfuryl thioacetate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

W316202-100G-K W316202-1KG-K
SIALW316202-100G-KEA 125 EUR
SIALW316202-100G-K SIALW316202-1KG-K
S-Furfuryl thioacetate, Sigma-Aldrich®
S-Furfuryl thioacetate
Formula: C₇H₈O₂S
MDL Number: MFCD00010083
CAS Number: 13678-68-7
EINECS: 237-173-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR