2-(Tributylstannyl)furan, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
414506-25ML
414506-5ML
SIAL414506-25MLEA
127
EUR
SIAL414506-25ML
SIAL414506-5ML
2-(Tributylstannyl)furan, Sigma-Aldrich®
2-(Tributylstannyl)furan
Formula:
C₁₆H₃₀OSn |
MDL Number:
MFCD00192512 CAS Number: 118486-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...