Methyl-4-acetyl-1,2,5-trimethylpyrrole-3-carboxylate

Supplier: Thermo Scientific

SP00012.250 SP00012.1
MAYBSP00012.250EA 0 EUR
MAYBSP00012.250 MAYBSP00012.1
Methyl-4-acetyl-1,2,5-trimethylpyrrole-3-carboxylate
Methyl 4-acetyl-1,2,5-trimethylpyrrole-3-carboxylate
Formula: C₁₁H₁₅NO₃
CAS Number: 175276-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR