5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulphonic acid disodium salt hydrate ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 3-(2-Pyridyl)-5,6-bis(4-sulphophenyl)-1,2,4-triazine disodium salt hydrate, 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulphonic acid disodium salt hydrate, PDT disulphonate hydrate, FerroZine(TM) Iron reagent disodium salt hydrate
L10607.03 L10607.06 L10607.PG
L10607.03EA 33.3 EUR
L10607.03 L10607.06 L10607.PG
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulphonic acid disodium salt hydrate ≥98%
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulphonic acid disodium salt hydrate
Purity: 98% CAS No.: 28048-33-1 Molecular Formula: C20H12N4Na2O6S2.xH2O Formula Weight: 514.46(anhy) Beilstein Reference: 5222087 MDL No.: MFCD00717621
Formula: C₂₀H₁₂N₄Na₂O₆S₂·H₂O
MW: 514.44 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00717621
CAS Number: 28048-33-1
EINECS: 248-797-6