2,5-Dimethylpyrrole-3-carboxylic acid ≥97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2,5-Dimethyl-1H-pyrrole-3-carboxylic acid
B20635.03 B20635.06 B20635.14
B20635.03EA 96.3 EUR
B20635.03 B20635.06 B20635.14
2,5-Dimethylpyrrole-3-carboxylic acid ≥97%
2,5-Dimethylpyrrole-3-carboxylic acid
Purity: 97% CAS No.: 57338-76-8 Molecular Formula: C7H9NO2 Formula Weight: 139.16 Melting Point: 199-201° dec. MDL No.: MFCD00203874
Formula: C₇H₉NO₂
MW: 139.16 g/mol
Melting Pt: 199…201 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00203874
CAS Number: 57338-76-8