4-Bromo-N,N-diphenylaniline 99%
Supplier: Thermo Fisher Scientific
Warning
H60328.06 H60328.14
H60328.06EA 92.1 EUR
H60328.06 H60328.14
4-Bromo-N,N-diphenylaniline 99%
4-Bromo-N,N-diphenylaniline
Formula: C₁₈H₁₄BrN
Melting Pt: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD01851266
CAS Number: 36809-26-4