Perfluorodecanoic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

177741-25G 177741-5G
SIAL177741-25GEA 197 EUR
SIAL177741-25G SIAL177741-5G
Perfluorodecanoic acid, Sigma-Aldrich®
Perfluorodecanoic acid
Formula: C₁₀HF₁₉O₂
MDL Number: MFCD00004175
CAS Number: 335-76-2
EINECS: 206-400-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR