Manganese(II) chloride hydrate, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

529680-25G 529680-5G
SIAL529680-25GEA 282 EUR
SIAL529680-25G SIAL529680-5G
Manganese(II) chloride hydrate, Sigma-Aldrich®
Manganese (II) chloride hydrate
Formula: Cl₂Mn·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149793
CAS Number: 73913-06-1
EINECS: 231-869-6
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR