2,5-Dimethylpyrrole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,5-Dimethyl-1H-pyrrole
M-Clarity™ quality level = MQ100
D183601-25G D183601-5G
SIALD183601-25GEA 106 EUR
SIALD183601-25G SIALD183601-5G
2,5-Dimethylpyrrole, Sigma-Aldrich®
2,5-Dimethylpyrrole
Formula: C₆H₉N
MW: 95.1424 g/mol
MDL Number: MFCD00005223
CAS Number: 625-84-3
EINECS: 210-913-8