2-Fluoro-3-nitropyridine ≥98%
Supplier: Thermo Fisher Scientific
Warning
H61260.14 H61260.22
H61260.14EA 98.6 EUR
H61260.14 H61260.22
2-Fluoro-3-nitropyridine ≥98%
2-Fluoro-3-nitropyridine
Formula: C₅H₃FN₂O₂
Boiling Pt: 110 °C (10 mmHg)
Melting Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD03095068
CAS Number: 1480-87-1