cis-4-Isopropylcyclohexanecarboxylic acid ≥97%
Supplier: Thermo Fisher Scientific
Warning
H62594.MD H62594.03 H62594.06
H62594.MDEA 83.3 EUR
H62594.MD H62594.03 H62594.06
cis-4-Isopropylcyclohexanecarboxylic acid ≥97%
cis-4-Isopropylcyclohexanecarboxylic acid
Formula: C₁₀H₁₈O₂
Storage Temperature: Ambient
CAS Number: 7084-93-7
EINECS: 230-385-2