Tungsten, gauze woven from wire 40 mesh woven from 0.025mm (0.001in)
Supplier: Thermo Fisher Scientific
46412.FL 46412.GT
46412.FLEA 61.9 EUR
46412.FL 46412.GT
Tungsten, gauze woven from wire 40 mesh woven from 0.025mm (0.001in)
Tungsten
Formula: W
Boiling Pt: 5660 °C (1013 hPa)
Melting Pt: 3410 °C
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945