1,5-Diphenylcarbazone
Supplier: Thermo Fisher Scientific
H60336.06 H60336.14
H60336.06EA 144 EUR
H60336.06 H60336.14
1,5-Diphenylcarbazone
1,5-Diphenylcarbazone
Formula: C₁₃H₁₂N₄O
Boiling Pt: 440 °C (1013 hPa)
Melting Pt: 119…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00003024
CAS Number: 538-62-5
EINECS: 208-698-0
Merck Index: 12,03380