3,4-Dichloro-N-methylaniline
Supplier: Thermo Scientific
Synonyms: N1-Methyl-3,4-dichloroaniline
TL00244.1 TL00244.50
MAYBTL00244.1EA 0 EUR
MAYBTL00244.1 MAYBTL00244.50
3,4-Dichloro-N-methylaniline
3,4-Dichloro-N-methylaniline
Formula: C₇H₇Cl₂N
Boiling Pt: 142…145 °C (1013 hPa)
MDL Number: MFCD00052850
CAS Number: 40750-59-2
UN: 2810
ADR: 6.1,III