Thymol blue 0,04% aqueous solution
Supplier: Lovibond
Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
451650 451651
LOVI451650EA 23.1 EUR
LOVI451650 LOVI451651
Thymol blue 0,04% aqueous solution
Thymol blue
Formula: C₂₇H₃₀O₅S
MW: 466.6 g/mol
Melting Pt: 221
Storage Temperature: Ambient
MDL Number: MFCD00005869
CAS Number: 76-61-9
EINECS: 200-973-3
Merck Index: 12,09541