Gallic acid
Supplier: Cayman Chemical
Synonyms: 3,4,5-Trihydroxybenzoic acid
11846-50 11846-10 11846-100
CAYM11846-50EA 75 EUR
CAYM11846-50 CAYM11846-10 CAYM11846-100
Gallic acid
Gallic acid
Formula: C₇H₆O₅
MW: 170.12 g/mol
Boiling Pt: 501
Melting Pt: 251…252 °C
Density: 1.694 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002510
CAS Number: 149-91-7
EINECS: 205-749-9
Merck Index: 13,04366