4-Chlorophenethylamine 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-(4-Chlorophenyl)ethylamine
H63081.06 H63081.14 H63081.22
H63081.06EA 31.1 EUR
H63081.06 H63081.14 H63081.22
4-Chlorophenethylamine 97%
4-Chlorophenethylamine
Formula: C₈H₁₀ClN
MW: 155.63 g/mol
Boiling Pt: 60…65 °C (0.1 mmHg)
Density: 1.120
Flash Pt: 106 °C (223 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008191
CAS Number: 156-41-2
EINECS: 205-853-4