2,3,6-Trimethylphenol 95%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 3-Hydroxypseudocumene
H60489.14 H60489.22
H60489.14EA 37.8 EUR
H60489.14 H60489.22
2,3,6-Trimethylphenol 95%
2,3,6-Trimethylphenol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 215 °C
Melting Pt: 59…62 °C
Density: 0.940
Flash Pt: 100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002229
CAS Number: 2416-94-6
EINECS: 219-330-3
UN: 2430
ADR: 8 III