Tetracycline hydrochloride
Supplier: Cayman Chemical
Warning
14328-10 14328-25 14328-50
CAYM14328-10EA 48 EUR
CAYM14328-10 CAYM14328-25 CAYM14328-50
Tetracycline hydrochloride
Tetracycline hydrochloride
Formula: C₂₂H₂₅ClN₂O₈
MW: 480.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078142
CAS Number: 64-75-5
EINECS: 200-593-8