Chemical resistant gloves, Alto 405
Supplier: MAPA
34405098 34405099 34405090 34405177
MAPA34405098EA 4.2 EUR
MAPA34405098 MAPA34405099 MAPA34405090 MAPA34405177
Chemical resistant gloves, Alto 405
Gloves
Neoprene/latex gloves with flocked interior lining and fine touch for light chemical protection.
  • Tactile sensitivity due to the gloves reduced thickness
  • Cotton flock lining provides comfort and perspiration absorption
  • Good grip due to the non-slip embossing
Ideal for the automotive and mechanical industry, spray-painting, industrial cleaning, maintenance and for work with composite materials (resins).
Certifications: EN 388:2016 (2110X), EN ISO 374-1:2016 Type B (KMT), EN 421; Cat. III